Learn More
MYB/c-Myb Mouse, anti-Human, Clone: 2C4A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68542-1-IG-20UL
Description
The MYB/c-Myb antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYB/c-Myb. This antibody recognizes human antigen. The MYB/c-Myb antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| MYB/c-Myb | |
| 2C4A5 | |
| c myb, c myb_CDS, Cmyb, efg, MYB, Proto oncogene c Myb, Transcriptional activator Myb | |
| 20 μL | |
| 4602 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Flow Cytometry, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |