missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYB/c-Myb Mouse, anti-Human, Clone: 2C4A5, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MYB/c-Myb |
|---|---|
| Clone | 2C4A5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18028735
|
Proteintech
68542-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18057885
|
Proteintech
68542-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MYB/c-Myb antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYB/c-Myb. This antibody recognizes human antigen. The MYB/c-Myb antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| MYB/c-Myb | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| c myb, c myb_CDS, Cmyb, efg, MYB, Proto oncogene c Myb, Transcriptional activator Myb | |
| Primary |
| 2C4A5 | |
| Western Blot, Flow Cytometry, ELISA | |
| Mouse | |
| 4602 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title