Learn More
MYB/c-Myb Mouse, anti-Human, Clone: 2C4A5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68542-1-IG-150UL
 View more versions of this product
Product Code. 18057885
  • € 410.00 / 150µL
Description
The MYB/c-Myb antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYB/c-Myb. This antibody recognizes human antigen. The MYB/c-Myb antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
MYB/c-Myb
2C4A5
c myb, c myb_CDS, Cmyb, efg, MYB, Proto oncogene c Myb, Transcriptional activator Myb
150 μL
4602
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Flow Cytometry, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa