missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium avium subsp. paratuberculosis Antibody (6G11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54646-0.025mg
This item is not returnable.
View return policy
Description
Mycobacterium avium subsp. paratuberculosis Monoclonal specifically detects Mycobacterium avium subsp. paratuberculosis in Bacteria samples. It is validated for ELISA, Immunoprecipitation.
Specifications
| Mycobacterium avium subsp. paratuberculosis | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Mycobacterium paratuberculosis | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgM |
| ELISA, Immunoprecipitation, Immunofluorescence | |
| 6G11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RNF135 | |
| Gamma-irradiated whole MAP strain B4 cells | |
| 0.025 mg | |
| Virology Bacteria and Parasites | |
| 84282 | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction