missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium avium subsp. paratuberculosis Antibody (6G11), Novus Biologicals™
Mouse Monoclonal Antibody
€ 206.00 - € 498.00
Specifications
| Antigen | Mycobacterium avium subsp. paratuberculosis |
|---|---|
| Clone | 6G11 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 |
| Applications | ELISA, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18055026
|
Novus Biologicals
NBP2-54646 |
0.1 mg |
€ 498.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600387
|
Novus Biologicals
NBP2-54646-0.025mg |
0.025 mg |
€ 206.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mycobacterium avium subsp. paratuberculosis Monoclonal specifically detects Mycobacterium avium subsp. paratuberculosis in Bacteria samples. It is validated for ELISA, Immunoprecipitation.Specifications
| Mycobacterium avium subsp. paratuberculosis | |
| 1.0 mg/mL | |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Virology Bacteria and Parasites | |
| PBS with 0.05% Sodium Azide | |
| Gamma-irradiated whole MAP strain B4 cells | |
| Primary |
| 6G11 | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bacteria | |
| Mycobacterium paratuberculosis | |
| IgM | |
| Thiophilic Chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title