missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium avium subsp. paratuberculosis Antibody (6G11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54646
This item is not returnable.
View return policy
Description
Mycobacterium avium subsp. paratuberculosis Monoclonal specifically detects Mycobacterium avium subsp. paratuberculosis in Bacteria samples. It is validated for ELISA, Immunoprecipitation.
Specifications
| Mycobacterium avium subsp. paratuberculosis | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Mycobacterium paratuberculosis | |
| Gamma-irradiated whole MAP strain B4 cells | |
| 0.1 mg | |
| Virology Bacteria and Parasites | |
| Bacteria | |
| Purified |
| ELISA, Immunoprecipitation | |
| 6G11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction