missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium tuberculosis Antibody (BGN/1209/3875) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62769-0.025MG
This item is not returnable.
View return policy
Description
Mycobacterium tuberculosis Monoclonal specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| Mycobacterium tuberculosis | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| M. tuberculosis | |
| Recombinant 38 kDa antigen from Mycobacterium tuberculosis. | |
| 0.025 mg | |
| Primary | |
| Bacteria | |
| Purified |
| Western Blot, ELISA | |
| BGN/1209/3875 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| This product recognises the 38 kDa antigen from Mycobacterium tuberculosis, a peptide believed to be involved in the regulation of proteins necessary for virulence. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction