missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium tuberculosis Antibody (BGN/1209/3875) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 204.00 - € 499.00
Specifications
| Antigen | Mycobacterium tuberculosis |
|---|---|
| Clone | BGN/1209/3875 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18081616
|
Novus Biologicals
NB100-62769-0.025MG |
0.025 mg |
€ 204.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18049055
|
Novus Biologicals
NB100-62769 |
0.1 mg |
€ 499.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mycobacterium tuberculosis Monoclonal specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
| Mycobacterium tuberculosis | |
| 1.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Recombinant 38 kDa antigen from Mycobacterium tuberculosis. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| BGN/1209/3875 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| M. tuberculosis | |
| IgG2b | |
| Protein A purified | |
| This product recognises the 38 kDa antigen from Mycobacterium tuberculosis, a peptide believed to be involved in the regulation of proteins necessary for virulence. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title