missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E4B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66286-1-IG-150UL
This item is not returnable.
View return policy
Description
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
| MYL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08590, P09542, P16409 | |
| Myl3 | |
| MYL3 Fusion Protein Ag24535 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2E4B2 | |
| Unconjugated | |
| MYL3 | |
| Cardiac myosin light chain 1, CMH8, CMLC1, MLC1SB, MLC1V, MYL3, Myosin light chain 3, VLC1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100515755, 17897, 24585, 4634 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur