missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E4B2, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MYL3 |
|---|---|
| Clone | 2E4B2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16818025
|
Proteintech
66286-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808025
|
Proteintech
66286-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
| MYL3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Cardiac myosin light chain 1, CMH8, CMLC1, MLC1SB, MLC1V, MYL3, Myosin light chain 3, VLC1 | |
| Myl3 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2E4B2 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig, Rat | |
| P08590, P09542, P16409 | |
| 100515755, 17897, 24585, 4634 | |
| MYL3 Fusion Protein Ag24535 | |
| Primary | |
| -20°C | |
| MYL3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title