missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Antibody (SR1600), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21922-50ul
This item is not returnable.
View return policy
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| Myogenin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| SR1600 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunohistochemistry 1:50-1:200 | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| A synthesized peptide derived from human Myogenin (Uniprot #: P15173) | |
| 50 μg | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| 4656 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction