missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Antibody (SR1600), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | Myogenin |
|---|---|
| Clone | SR1600 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18609933
|
Novus Biologicals
NBP3-21922-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664044
|
Novus Biologicals
NBP3-21922-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| Myogenin | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| A synthesized peptide derived from human Myogenin (Uniprot #: P15173) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1600 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4656 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title