missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYSM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20078-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to MYSM1.
MYSM1is a metalloprotease that specifically deubiquitinates monoubiquitinated histone H2A, a specific tag for epigenetic transcriptional repression, thereby acting as a coactivator. Preferentially deubiquitinates monoubiquitinated H2A in hyperacetylated nucleosomes. Deubiquitination of histone H2A leads to facilitate the phosphorylation and dissociation of histone H1 from the nucleosome. Acts as a coactivator by participating in the initiation and elongation steps of androgen receptor (AR)-induced gene activation.Specifications
| MYSM1 | |
| Polyclonal | |
| Unconjugated | |
| MYSM1 | |
| 2A DUB, 2ADUB, KIAA1915, MYSM1, RP4 592A1.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114803 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5VVJ2 | |
| MYSM1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction