missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYSM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Tekniske data
| Antigen | MYSM1 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|
16819564
|
Proteintech
20078-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809564
|
Proteintech
20078-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
The antibody is specific to MYSM1.
MYSM1is a metalloprotease that specifically deubiquitinates monoubiquitinated histone H2A, a specific tag for epigenetic transcriptional repression, thereby acting as a coactivator. Preferentially deubiquitinates monoubiquitinated H2A in hyperacetylated nucleosomes. Deubiquitination of histone H2A leads to facilitate the phosphorylation and dissociation of histone H1 from the nucleosome. Acts as a coactivator by participating in the initiation and elongation steps of androgen receptor (AR)-induced gene activation.Tekniske data
| MYSM1 | |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5VVJ2 | |
| 114803 | |
| Peptide | |
| Primary | |
| -20°C | |
| MYSM1 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 2A DUB, 2ADUB, KIAA1915, MYSM1, RP4 592A1.1 | |
| MYSM1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel