Learn More
MYST1 Antibody (7E11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26184-100ul
 View more versions of this product
Product Code. 30026707
  • € 533.00 / 100µL
Description
MYST1 Monoclonal antibody specifically detects MYST1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MYST1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunocytochemistry
7E11
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1
A synthesized peptide derived from Human MYST1 [UniProt Q9H7Z6]
100 μL
Chromatin Research, DNA Repair
84148
Store at -20 to -70C. Avoid freeze-thaw cycles.