missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYST1 Antibody (7E11), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | MYST1 |
|---|---|
| Clone | 7E11 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30038719
|
Novus Biologicals
NBP3-26184-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30026707
|
Novus Biologicals
NBP3-26184-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYST1 Monoclonal antibody specifically detects MYST1 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| MYST1 | |
| ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1 | |
| A synthesized peptide derived from Human MYST1 [UniProt Q9H7Z6] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7E11 | |
| ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, DNA Repair | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 84148 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title