missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYST1 Antibody (7E11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26184-50ul
This item is not returnable.
View return policy
Description
MYST1 Monoclonal antibody specifically detects MYST1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| MYST1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry | |
| 7E11 | |
| ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1 | |
| A synthesized peptide derived from Human MYST1 [UniProt Q9H7Z6] | |
| 50 μL | |
| Chromatin Research, DNA Repair | |
| 84148 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction