Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
Brand: Macherey-Nagel™ 70231.3
Additional Details : Weight : 0.01000kg
Description
- Ready assembled crimp closures and plain crimp caps N 11
- Septa are assembled, caps are ready-to-use.
- Septa are shown for illustration purposes only, images upside down.
- Packs of 100 closures each
Specifications
Blue | |
Natural rubber / Butyl red orange/ TEF colorless | |
1 mm |
Aluminum, Butyl Rubber, Rubber, TEF | |
100 Pk. | |
Crimp Cap |