missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,4-diaminobutane, 97+%

Product Code. 15402988
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15402988 5 mL 5mL
15412988 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402988 Supplier Thermo Scientific Alfa Aesar Supplier No. H56499.AA
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 68076-36-8 | C9H20N2O2 | 188.271 g/mol

N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68076-36-8
Molecular Formula C9H20N2O2
Molecular Weight (g/mol) 188.271
MDL Number MFCD00210019
InChI Key ZFQWJXFJJZUVPI-UHFFFAOYSA-N
Synonym tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane
PubChem CID 4351
IUPAC Name tert-butyl N-(4-aminobutyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCN

Specifications

Linear Formula (CH3)3COCONH(CH2)4NH2
Refractive Index 1.46
UN Number UN2735
Beilstein 1937878
Density 0.984
Quantity 5 mL
Flash Point 109°C (228°F)
Formula Weight 188.27
Percent Purity 98%
Chemical Name or Material N-Boc-1,4-diaminobutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.