Learn More
N-cadherin Mouse, anti-Human, Clone: 2G7C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68532-1-IG-150UL
Description
The N-cadherin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-cadherin. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The N-cadherin antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| N-cadherin | |
| 2G7C2 | |
| Cadherin 2, CD325, CDH2, CDHN, CDw325, N cadherin, NCAD, Neural cadherin | |
| 150 μL | |
| 1000 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse, Rat, Rabbit | |
| 150 kDa | |
| IgG2b |