Learn More
N-cadherin Mouse, anti-Human, Clone: 2G7C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68532-1-IG-150UL
 View more versions of this product
Product Code. 18065285
  • € 410.00 / 150µL
Description
The N-cadherin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-cadherin. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The N-cadherin antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
N-cadherin
2G7C2
Cadherin 2, CD325, CDH2, CDHN, CDw325, N cadherin, NCAD, Neural cadherin
150 μL
1000
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
1000 μg/mL
Mouse
Primary
Human, Pig, Mouse, Rat, Rabbit
150 kDa
IgG2b