Learn More
N-cadherin Mouse, anti-Human, Clone: 2G7C2, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | N-cadherin |
|---|---|
| Clone | 2G7C2 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18064785
|
Proteintech
68532-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18065285
|
Proteintech
68532-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The N-cadherin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-cadherin. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The N-cadherin antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| N-cadherin | |
| 1000 μg/mL | |
| Liquid | |
| Human, Pig, Mouse, Rat, Rabbit | |
| Cadherin 2, CD325, CDH2, CDHN, CDw325, N cadherin, NCAD, Neural cadherin | |
| Primary |
| 2G7C2 | |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| Mouse | |
| 1000 | |
| IgG2b | |
| Store at -20°C. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.