Learn More
N1-(3-acetyl-4-hydroxyphenyl)acetamide, Thermo Scientific™
Product Code. 10096412
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10096412 1 g 1g
10230205 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10096412 Supplier Thermo Scientific Maybridge Supplier No. BTB10078DA
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Chemical Identifiers
CAS 7298-67-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
InChI Key DIQSYMRVTOVKQT-UHFFFAOYSA-N
Synonym n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide
PubChem CID 81720
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)acetamide
SMILES CC(=O)C1=C(C=CC(=C1)NC(=O)C)O
Specifications
Assay Percent Range 97%
Quantity 1 g
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material N1-(3-Acetyl-4-hydroxyphenyl)acetamide