Learn More
NaCl 0.9% Rinse, B Braun™
Product Code. 10258674
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
10 bottles
6 bottles
Product Code. 10258674
Brand: B Braun 3570160
Edge
Use B Braun™ NaCl 0.9% Rinse for all general irrigation, washing, rinsing and dilution purposes which permit use of a sterile, nonpyrogenic electrolyte solution.
  • Sterile
  • Nonpyrogenic
  • Isotonic
  • Electrolyte solution
  • Contains no bacteriostatic or antimicrobial agents
  • No natural rubber latex, DEHP or PVC
Specifications
Quantity 1000 mL