missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanos2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76370-0.025mg
This item is not returnable.
View return policy
Description
Nanos2 Polyclonal specifically detects Nanos2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Nanos2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| nanos homolog 2, nanos homolog 2 (Drosophila), NOS2NOS-2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 339345 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2-4 μg/mL, ELISA 1:100-1:2000 | |
| P60321 | |
| NANOS2 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of the human Nanos2. The immunogen is located within the last 50 amino acids of Nanos2. Amino Acid Squence: QSLYRRSGRNSAGRR | |
| 0.025 mg | |
| Primary | |
| This Nanos2 antibody will not cross-react with either Nanos 1 or Nanos3. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction