missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanos2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 168.00
Specifications
| Antigen | Nanos2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2-4 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Nanos2 Polyclonal specifically detects Nanos2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| Nanos2 | |
| Western Blot 2-4 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P60321 | |
| 339345 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of the human Nanos2. The immunogen is located within the last 50 amino acids of Nanos2. Amino Acid Squence: QSLYRRSGRNSAGRR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| nanos homolog 2, nanos homolog 2 (Drosophila), NOS2NOS-2 | |
| NANOS2 | |
| IgG | |
| Affinity Purified | |
| This Nanos2 antibody will not cross-react with either Nanos 1 or Nanos3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title