Learn More
Thermo Scientific™ 20mm Crimp Seals with Septa for Headspace Vials
Product Code. 12400298
Click to view available options
Septum:
Aluminum Foil/White High Temp. Silicone
Clear PTFE/gray butyl
SepCap Integral molded Polyethylene
Product Type:
Crimp Cap
Pressure Release
Unit Size:
Pack of 100
Pack of 250
Product Code. 12400298
Brand: Thermo Scientific™ C402043A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Minimize the risk of contamination with quick, convenient Thermo Scientific™ National 20mm Crimp Seals with Septa.
Minimize the risk of contamination with quick, convenient Thermo Scientific™ National 20mm Crimp Seals with Septa.
20mm Crimp seals with prefitted septa for headspace vials
  • Seals with pre-inserted septa are convenient and minimize contamination due to handling
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
  • PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
  • Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
Ordering Information:
Septa and crimp seals also sold separately.
Note:
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
TRUSTED_SUSTAINABILITY
Specifications
Description Clear PTFE/Gray Butyl septa
Material Butyl Rubber, PTFE, Magnetic Steel
Septum Clear PTFE/gray butyl
Product Line National
Closure Size 20 mm
For Use With (Equipment) Headspace Vials
Product Type Crimp Cap