Learn More
Thermo Scientific™ National Unassembled Target LoVial™ Wide-Opening Vial Kits
Click to view available options
Volume (Metric):
2 mL
4 mL
Septum:
PTFE/RR w/C4011-630 Insert
PTFE/Red Rubber
PTFE/Silicone
Color:
Amber
Clear
Neck Style:
Standard
Wide
Unit Size:
Pack of 100
Pack of 500
Description
Keep all vials and compatible crimp seals handy and dust-free with Thermo Scientific™ National Unassembled Target LoVial Wide-Opening Vial Kits.
- Two-compartment benchtop storage tray
- Include matched quantities of vials and silver aluminum seals with prefitted septa
Specifications
Specifications
| Closure Color | Silver |
| Description | Amber Glass; ID patchSilver Seal, PTFE/Red Rubber Septum; 4mL |
| Color | Amber |
| Material | PTFE, Rubber |
| Volume (Metric) | 4 mL |
| Septum | PTFE/Red Rubber |
| Neck Style | Wide |
| Closure Material | Aluminum |
| Marking Spot | Yes |