Learn More
Fisherbrand™ Natural HDPE Wide Mouth Oblong Bottles without Caps
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Pack of 116
Pack of 160
Description
- Natural HDPE Wide Mouth Oblong Bottles offer a space saving design for the workbench.
- Bottles feature a wide mouth opening, ideal for sampling solids and powders.
- HDPE provides an excellent moisture barrier and are impact resistant.
- Ideal for high-speed filling, demanding pharmaceutical specifications and rough handling when shipping.
- HDPE provides moisture barrier and are impact resistant
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Natural |
| Diameter (Metric) Outer Neck | 39 mm |
| Diameter (Metric) Inner Neck | 35 mm |
| Material | HDPE |
| Quantity | 160 Pk. |
| Neck Finish | 43-400 |
| Closure Included | No |
| Mouth | Wide |
| Height (Metric) | 149 mm |
| Show More |