missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20175-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Several alternatively spliced transcript variants have been found for this gene.Specifications
| NCOA5 | |
| Polyclonal | |
| Unconjugated | |
| NCOA5 | |
| CIA, KIAA1637, NCoA 5, NCOA5, nuclear receptor coactivator 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 228869, 296372, 57727 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Flow Cytometry | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91W39, Q9HCD5 | |
| NCOA5 | |
| NCOA5 Fusion Protein Ag14014 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction