missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | NCOA5 |
|---|---|
| Concentration | 0.31 mg/mL |
| Applications | Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16839684
|
Proteintech
20175-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16829684
|
Proteintech
20175-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Several alternatively spliced transcript variants have been found for this gene.Specifications
| NCOA5 | |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q91W39, Q9HCD5 | |
| 228869, 296372, 57727 | |
| NCOA5 Fusion Protein Ag14014 | |
| Primary | |
| -20°C | |
| NCOA5 |
| 0.31 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CIA, KIAA1637, NCoA 5, NCOA5, nuclear receptor coactivator 5 | |
| NCOA5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title