missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA4L2 Mouse anti-Human, Clone: 1G1H10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66050-1-IG-150UL
This item is not returnable.
View return policy
Description
NDUFA4L2 is a protein coding gene.Specifications
| NDUFA4L2 | |
| Monoclonal | |
| 1.06 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NRX3 | |
| NDUFA4L2 | |
| NDUFA4L2 Fusion Protein Ag9233 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1G1H10 | |
| Unconjugated | |
| NDUFA4L2 | |
| NDUFA4L2, NUOMS | |
| Mouse | |
| Protein A | |
| RUO | |
| 56901 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction