missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA4L2 Mouse anti-Human, Clone: 1G1H10, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | NDUFA4L2 |
|---|---|
| Clone | 1G1H10 |
| Concentration | 1.06 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16867575
|
Proteintech
66050-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16857575
|
Proteintech
66050-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFA4L2 is a protein coding gene.Specifications
| NDUFA4L2 | |
| 1.06 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| NDUFA4L2, NUOMS | |
| NDUFA4L2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1G1H10 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NRX3 | |
| 56901 | |
| NDUFA4L2 Fusion Protein Ag9233 | |
| Primary | |
| -20°C | |
| NDUFA4L2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title