Learn More
NDUFA4L2 Mouse anti-Human, Clone: 1G1H10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66050-1-IG-20UL
 View more versions of this product
Product Code. 16867575
  • € 145.00 / 20µL
Description
NDUFA4L2 is a protein coding gene.
Specifications
Specifications
NDUFA4L2
Monoclonal
1.06 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NRX3
NDUFA4L2
NDUFA4L2 Fusion Protein Ag9233
20 μL
Primary
Human
Antibody
IgG2a
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
1G1H10
Unconjugated
NDUFA4L2
NDUFA4L2, NUOMS
Mouse
Protein A
RUO
56901
-20°C