missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27931-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
| NDUFB4 | |
| Polyclonal | |
| Unconjugated | |
| NDUFB4 | |
| B15, CI B15, Complex I B15, NDUFB4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4710, 68194 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95168, Q9CQC7 | |
| NDUFB4 | |
| NDUFB4 Fusion Protein Ag27484 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction