Learn More
Invitrogen™ NDUFC2 Recombinant Rabbit Monoclonal Antibody (24GB505)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557105
 View more versions of this product
Product Code. 30275719
  • € 500.00 / 100µL
Description
NDUFC2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NDUFC2
Recombinant Monoclonal
Unconjugated
NDUFS2
14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 1810004I06Rik; 2010300P09Rik; B14.5b; CI-B14.5b; CI-B145b; complex I subunit B14.5b; complex I-B14.5b; Complex I-B145b; G1; HLC1; HLC-1; Human lung cancer oncogene 1 protein; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 subunit C2; NADH:ubiquinone oxidoreductase subunit C2; NADHDH2; NADH-ubiquinone oxidoreductase subunit B14.5b; Ndufc2; R75094
Rabbit
Affinity chromatography
RUO
4718, 4720
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB505
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75306, O95298
NDUFC2, NDUFS2
A synthesized peptide derived from human NDUFC2 (aa 1-119).