missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFC2 Recombinant Rabbit Monoclonal Antibody (24GB505)

Rabbit Recombinant Monoclonal Antibody
€ 146.00 - € 500.00
Specifications
| Antigen | NDUFC2 |
|---|---|
| Clone | 24GB505 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275719
![]() |
Invitrogen™
MA557105 |
100 μL |
€ 500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275654
|
Invitrogen™
MA557106 |
20 μL |
€ 146.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFC2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
| NDUFC2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| NDUFS2 | |
| 14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 1810004I06Rik; 2010300P09Rik; B14.5b; CI-B14.5b; CI-B145b; complex I subunit B14.5b; complex I-B14.5b; Complex I-B145b; G1; HLC1; HLC-1; Human lung cancer oncogene 1 protein; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 subunit C2; NADH:ubiquinone oxidoreductase subunit C2; NADHDH2; NADH-ubiquinone oxidoreductase subunit B14.5b; Ndufc2; R75094 | |
| NDUFC2, NDUFS2 | |
| Primary | |
| Affinity chromatography |
| 24GB505 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75306, O95298 | |
| 4718, 4720 | |
| A synthesized peptide derived from human NDUFC2 (aa 1-119). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title