missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurocan Antibody (SR1121), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22319-100ul
This item is not returnable.
View return policy
Description
Neurocan Monoclonal antibody specifically detects Neurocan in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| Neurocan | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1121 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Chondroitin sulfate proteoglycan 3, chondroitin sulfate proteoglycan 3 (neurocan), CSPG3neurocan core protein, FLJ44681, NEUR, neurocan, neurocan proteoglycan | |
| A synthesized peptide derived from human Neurocan (Uniprot #: O14594) | |
| 100 μg | |
| Cell Biology | |
| 1463 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction