missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurocan Antibody (SR1121), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Neurocan |
|---|---|
| Clone | SR1121 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649094
|
Novus Biologicals
NBP3-22319-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648884
|
Novus Biologicals
NBP3-22319-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurocan Monoclonal antibody specifically detects Neurocan in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| Neurocan | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Chondroitin sulfate proteoglycan 3, chondroitin sulfate proteoglycan 3 (neurocan), CSPG3neurocan core protein, FLJ44681, NEUR, neurocan, neurocan proteoglycan | |
| A synthesized peptide derived from human Neurocan (Uniprot #: O14594) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1121 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1463 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title