missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21354-25ul
This item is not returnable.
View return policy
Description
Neurogenin-1 Polyclonal antibody specifically detects Neurogenin-1 in Human samples. It is validated for Immunofluorescence
Specifications
| Neurogenin-1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PNISRASEVPGAQDDEQERRRRRGRTRVRSEALLHSLRRS | |
| 25 μg | |
| Neuroscience | |
| 4762 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur