missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 425.25 - € 668.85
Specifications
| Antigen | Neurogenin-1 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600554
|
Novus Biologicals
NBP3-21354-25ul |
25 μg |
€ 450.00 € 425.25 / 25µL Save € 24.75 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662655
|
Novus Biologicals
NBP3-21354-100ul |
100 μg |
€ 708.00 € 668.85 / 100µL Save € 39.15 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenin-1 Polyclonal antibody specifically detects Neurogenin-1 in Human samples. It is validated for ImmunofluorescenceSpecifications
| Neurogenin-1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, pH 7.2, 40% glycerol | |
| 4762 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PNISRASEVPGAQDDEQERRRRRGRTRVRSEALLHSLRRS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title