missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21354-100ul
This item is not returnable.
View return policy
Description
Neurogenin-1 Polyclonal antibody specifically detects Neurogenin-1 in Human samples. It is validated for Immunofluorescence
Specifications
| Neurogenin-1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PNISRASEVPGAQDDEQERRRRRGRTRVRSEALLHSLRRS | |
| 100 μg | |
| Neuroscience | |
| 4762 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction