Learn More
NeutraKine™IL-12 p35 Mouse, anti-Human, Clone: 2G11G6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69029-1-IG-25UG
 View more versions of this product
Product Code. 18009445
  • € 149.00 / 25µg
Description
The NeutraKine™IL-12 p35 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™IL-12 p35. This antibody recognizes human antigen. The NeutraKine™IL-12 p35 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
Specifications
NeutraKine™IL-12 p35
2G11G6
Mouse
Primary
Human
150 kDa
IgG2b
ELISA, Neutralization
CLMF, CLMF p35, IL 12 subunit p35, IL 12A, IL-12 p35, IL12A, Interleukin 12 subunit alpha, NFSK, NKSF1, P35
25 μg
3592
Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.