Learn More
NeutraKine™IL-12 p35 Mouse, anti-Human, Clone: 2G11G6, Proteintech
Mouse Monoclonal Antibody
€ 149.00 - € 319.00
Specifications
| Antigen | NeutraKine™IL-12 p35 |
|---|---|
| Clone | 2G11G6 |
| Applications | ELISA, Neutralization |
| Form | Liquid |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18009445
|
Proteintech
69029-1-IG-25UG |
25 μg |
€ 149.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18048755
|
Proteintech
69029-1-IG-100UG |
100 μg |
€ 319.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™IL-12 p35 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™IL-12 p35. This antibody recognizes human antigen. The NeutraKine™IL-12 p35 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
| NeutraKine™IL-12 p35 | |
| ELISA, Neutralization | |
| Mouse | |
| 3592 | |
| IgG2b | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| 2G11G6 | |
| Liquid | |
| Human | |
| CLMF, CLMF p35, IL 12 subunit p35, IL 12A, IL-12 p35, IL12A, Interleukin 12 subunit alpha, NFSK, NKSF1, P35 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.