Learn More
NeutraKine™ IL-6 Mouse anti-Human, Unconjugated, Clone: 4G3E6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69001-1-IG-100UG
 View more versions of this product
Product Code. 16850406
  • € 319.00 / 100µg
Description
The NeutraKine™ IL-6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-6. This antibody recognizes human antigen. The NeutraKine™ IL-6 antibody has been validated for the following applications: FC, IHC, Neutralization, ELISA analysis.
Specifications
Specifications
NeutraKine™ IL-6
Monoclonal
Unconjugated
100 μg
3569
IgG1
Neutralization, Immunohistochemistry, Flow Cytometry
4G3E6