Learn More
NeutraKine™ IL-6 Mouse anti-Human, Unconjugated, Clone: 4G3E6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69001-1-IG-100UG
Description
The NeutraKine™ IL-6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-6. This antibody recognizes human antigen. The NeutraKine™ IL-6 antibody has been validated for the following applications: FC, IHC, Neutralization, ELISA analysis.
Specifications
| NeutraKine™ IL-6 | |
| Monoclonal | |
| Unconjugated | |
| 100 μg | |
| 3569 | |
| IgG1 |
| Neutralization, Immunohistochemistry, Flow Cytometry | |
| 4G3E6 | |
| Mouse | |
| Primary | |
| Human |