Learn More
NeutraKine™ IL-6 Mouse anti-Human, Unconjugated, Clone: 4G3E6, Proteintech
Mouse Monoclonal Antibody
€ 149.00 - € 319.00
Specifications
| Antigen | NeutraKine™ IL-6 |
|---|---|
| Clone | 4G3E6 |
| Applications | Neutralization, Immunohistochemistry, Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16860406
|
Proteintech
69001-1-IG-25UG |
25 μg |
€ 149.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16850406
|
Proteintech
69001-1-IG-100UG |
100 μg |
€ 319.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™ IL-6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-6. This antibody recognizes human antigen. The NeutraKine™ IL-6 antibody has been validated for the following applications: FC, IHC, Neutralization, ELISA analysis.
Specifications
| NeutraKine™ IL-6 | |
| Neutralization, Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| Human | |
| IgG1 |
| 4G3E6 | |
| Monoclonal | |
| Mouse | |
| 3569 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.