Learn More
NFATC2/NFAT1 Antibody (25A10.D6.D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand:  Novus Biologicals NB300-504-0.025mg
 View more versions of this product
Product Code. 18594580
  • € 213.00 / 25µg
Description
NFATC2/NFAT1 Monoclonal specifically detects NFATC2/NFAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays, Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NFATC2/NFAT1
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:2000, Immunohistochemistry-Frozen 1:50 - 1:2000, Gel Super Shift Assays 1:1 - 1:100, Sandwich ELISA 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1-3 ul
NFATC2
Synthetic peptide corresponding to residues A(51) I S S P S G L A Y P D D V L D Y G L(69) of mouse NFATc2-A, B and C isoforms.
Protein G purified
RUO
Primary
Detects nuclear factor of activated T-cells (NFAT). This does not cross react with NFAT2 (NFATc, NFATc1).
Store at -20°C. Avoid freeze/thaw cycles.
IgG1
Western Blot
25A10.D6.D2
Unconjugated
NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1
Mouse
140 kDa
0.025 mg
Adaptive Immunity, Immunology, Transcription Factors and Regulators, Wnt Signaling Pathway
4773