missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC2/NFAT1 Antibody (25A10.D6.D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
€ 213.00
Specifications
| Antigen | NFATC2/NFAT1 |
|---|---|
| Clone | 25A10.D6.D2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:2000, Immunohistochemistry-Frozen 1:50 - 1:2000, Gel Super Shift Assays 1:1 - 1:100, Sandwich ELISA 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1-3 ul |
| Applications | Western Blot |
Description
NFATC2/NFAT1 Monoclonal specifically detects NFATC2/NFAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays, Sandwich ELISA.Specifications
| NFATC2/NFAT1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology, Transcription Factors and Regulators, Wnt Signaling Pathway | |
| NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1 | |
| NFATC2 | |
| IgG1 | |
| Protein G purified | |
| Detects nuclear factor of activated T-cells (NFAT). This does not cross react with NFAT2 (NFATc, NFATc1). |
| 25A10.D6.D2 | |
| Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:2000, Immunohistochemistry-Frozen 1:50 - 1:2000, Gel Super Shift Assays 1:1 - 1:100, Sandwich ELISA 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 4773 | |
| Synthetic peptide corresponding to residues A(51) I S S P S G L A Y P D D V L D Y G L(69) of mouse NFATc2-A, B and C isoforms. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 140 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title