missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Alloy 800DS, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Alloy 800DS, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Aluminium/Titanium/Copper/Molybdenum/Cobalt/Manganese/Silicon/Chromium/Nickel |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon 0.08 %/Aluminium 0.083 %/Titanium 0.17 %/Copper 0.3 %/Molybdenum 0.3 %/Cobalt 0.5 %/Manganese 1.04 %/Silicon 1.98 %/Chromium 18.64 %/Nickel 36.57 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction