missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, PE16, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, PE16, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Oxygen/Boron/Nitrogen/Phosphorus/Vanadium/Zirconium/Tungsten/Niobium/Carbon/Copper/Manganese/Silicon/Cobalt/Titanium/Aluminium/Molybdenum/Chromium/Iron/Nickel |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Oxygen 0.0006 %/Boron 0.0028 %/Nitrogen 0.006 %/Phosphorus 0.006 %/Vanadium 0.018 %/Zirconium 0.019 %/Tungsten 0.038 %/Niobium 0.04 %/Carbon 0.052 %/Copper 0.053 %/Manganese 0.122 %/Silicon 0.13 %/Cobalt 0.77 %/Titanium 1.18 %/Aluminium 1.2 %/Molybdenum 3.29 %/Chromium 16.6 %/Iron 34.4 %/Nickel 42.9 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction