missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, High Purity Carbonyl Nickel-High, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, High Purity Carbonyl Nickel-High, Ø32x19mm
Specifications
Specifications
| Chemical Name or Material | Manganese/Cadmium/Thallium/Tin/Selenium/Cobalt/Zinc/Tellurium/Lead/Silver/Antimony/Bismuth/Copper/Carbon/Arsenic/Phosphorus/Aluminium/Iron/Vanadium/Beryllium/Molybdenum/Chromium/Nitrogen/Magnesium/Titanium/Oxygen/Gallium/Boron/Calcium/Nickel |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Manganese 0.00018 %/Cadmium 0.0005 %/Thallium 0.00058 %/Tin 0.00062 %/Selenium 0.00065 %/Cobalt 0.0008 %/Zinc 0.00081 %/Tellurium 0.00089 %/Lead 0.00093 %/Silver 0.00109 %/Antimony 0.0011 %/Bismuth 0.00111 %/Copper 0.0017 %/Carbon 0.0022 %/Arsenic 0.0028 %/Phosphorus 0.0034 %/Aluminium 0.005 %/Iron 0.0099 %/Vanadium (<0.00005) %/Beryllium (<0.0001) %/Molybdenum (<0.0001) %/Chromium (0.0001) %/Nitrogen (0.0001) %/Magnesium (0.0006) %/Titanium (0.0006) %/Oxygen (0.0019) %/Gallium <0.00005 %/Boron <0.0005 %/Calcium <0.0010 %/Nickel rem % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction